
9,10-Anthraquinone for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:84-65-1
:
Formula:C₁₄H₈O₂
MDL Number:MFCD00001188
:208.22 g/mol
Merck Index:13,00692
Boiling Pt:380 °C (1013 hPa)
Melting Pt:165…169 °C
Density:1.419…1.438 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:185 °C
EINECS:201-549-0
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Solubility:- 0.001 g/l (20 °C)
- Melting Point:282 - 285 °C
- Molar Mass:208.22 g/mol
- Bulk Density:600 kg/m3
- Boiling Point:379 - 381 °C
- Vapor Pressure:1.33 hPa (190 °C)
- Flash Point:185 °C
- Ignition Point:650 °C
- Synonyms:9,10-Dihydro-9,10-anthracenedione
- Hill Formula:C14H8O2