Order Entry
Norway
ContactUsLinkComponent
δ-BHC
Toxic
Health
Environment
δ-BHC
Catalog # 48495.
Supplier:  Merck
CAS Number:  
undefined
Toxic
Health
Environment
δ-BHC
Catalog # 48495.
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
 :290.83 g/mol
Boiling Pt:330 °C (1013 hPa)
Melting Pt:73…75 °C
Density:1.85 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:206-272-9
UN:2761
ADR:6.1,III
 :GHS06
 :GHS08
 :GHS09
CAS Number:319-86-8
Formula:C₆H₆Cl₆