


(1α,2α,3α,4β,5α,6β)-1,2,3,4,5,6-Heksaklorosykloheksan
Catalog # EHERC14074000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pakketype:Glass bottle for solids
- Fp:50 mg
- Seq:2370
Specifications
About this item
:
:290.83 g/mol
Boiling Pt:330 °C (1013 hPa)
Melting Pt:73…75 °C
Density:1,85 g/cm³ (20 °C)
Storage Temperature:Romtemperatur
EINECS:206-272-9
UN:2761
ADR:6.1,III
:GHS06
:GHS08
:GHS09
CAS Number:319-86-8
Formula:C₆H₆Cl₆