Order Entry
Norway
ContactUsLinkComponent
Benzotriazole ≥98%, TECHNICAL
Benzotriazole ≥98%, TECHNICAL
  21841.292
 :  
undefined
Benzotriazole ≥98%, TECHNICAL
  21841.292
 :  

 

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

 

 

  • Assay:
    Min. 98 %

 

 

 :
 :119.13 g/mol
 :13,01109
 :350 °C (1013 hPa)
 :99 °C
 :1.34 g/cm³ (20 °C)
 :170 °C (closed cup)
 :Ambient
 :202-394-1
 :2811
 :6.1,III
 :GHS09
 :GHS07
 :95-14-7
 :C₆H₅N₃
 :MFCD00005699