Order Entry
Norway
ContactUsLinkComponent
Benzotriazol
Flammable
Irritant
Benzotriazol
Katalog # MOLE35105688-500G
Supplier:  Molekula
CAS Number:  
undefined
Flammable
Irritant
Benzotriazol
Katalog # MOLE35105688-500G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pakketype:
    Glass bottle for solids
  • Fp:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :119.13 g/mol
Merck Index:13,01109
Boiling Pt:350 °C (1013 hPa)
Melting Pt:99 °C
Density:1,34 g/cm³ (20 °C)
Flash Pt:170 °C (closed cup)
Storage Temperature:Romtemperatur
EINECS:202-394-1
UN:2811
ADR:6.1,III
 :GHS02
 :GHS07
CAS Number:95-14-7
Formula:C₆H₅N₃
MDL Number:MFCD00005699