7-Hydroxycoumarin 99%
Supplier: Thermo Fisher Scientific
Om dette produktet
Synonymer: Umbelliferone, 7-Hydroxy-2H-chromen-2-one, 7-Hydroxy-2-benzopyrone
- CAS Number:93-35-6
- Formel:C₉H₆O₃
- MDL-nummer:MFCD00006878
- Molecular Weight:162.14 g/mol
- Merck-indeksen:12,09981
- Kokende Pt:382 °C (1013 hPa)
- Molten Pt:228…234 °C
- Lagringstemperatur:Ambient
- EINECS:202-240-3
2 Alternativer tilgjenglige nedenfor
Product Details & Documents
Synonymer: Umbelliferone, 7-Hydroxy-2H-chromen-2-one, 7-Hydroxy-2-benzopyrone
- CAS Number:93-35-6
- Formel:C₉H₆O₃
- MDL-nummer:MFCD00006878
- Molecular Weight:162.14 g/mol
- Merck-indeksen:12,09981
- Kokende Pt:382 °C (1013 hPa)
- Molten Pt:228…234 °C
- Lagringstemperatur:Ambient
- EINECS:202-240-3
Documents
ACRO12111
SDS
Specifications for Product Family
Spesifikasjonstestresultater
- Appearance:White to yellow to beige to light brown Powder or crystals and/or agglomerates
- Infrared spectrum:Conforms
- Melting point:228 to 234 °C
- HPLC:≥98.5 %
- Titration with NaOH:98.5 to 101.5 %



