Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,2',3,4',6'-Pentachlorobiphenyl (PCB No. 98)
2,2',3,4',6'-Pentachlorobiphenyl (PCB No. 98)
2,2',3,4',6'-Pentachlorobiphenyl (PCB No. 98)
2,2',3,4',6'-Pentachlorobiphenyl (PCB No. 98)
Catalog # PROOCIL-PCB-98-C
Supplier:
LGC Standards PROMOCHEM
CAS Number:
Expand image
2,2',3,4',6'-Pentachlorobiphenyl (PCB No. 98)
Catalog # PROOCIL-PCB-98-C
Supplier:
LGC Standards PROMOCHEM
CAS Number:
About this item
Product Details
Product Details