2,4-Dichlorobiphenyl (PCB No. 7)
Catalog # PROOCIL-PCB-7-C
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:223.1 g/mol
CAS Number:33284-50-3
Formula:C₁₂H₈Cl₂