Order Entry
Norway
ContactUsLinkComponent
2,2',3,3',4,5,6'-Heptachlorobiphenyl (PCB No. 174)
2,2',3,3',4,5,6'-Heptachlorobiphenyl (PCB No. 174)
Catalog # PROOCIL-PCB-174-C
CAS Number:  
2,2',3,3',4,5,6'-Heptachlorobiphenyl (PCB No. 174)
Catalog # PROOCIL-PCB-174-C
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :395.32 g/mol
CAS Number:38411-25-5
Formula:C₁₂H₃Cl₇