2,2',3,3',4,4',5-Heptachlorobiphenyl (PCB No. 170)
Catalog # PROOCIL-PCB-170-C
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:395.32 g/mol
UN:3432
ADR:9,II
CAS Number:35065-30-6
Formula:C₁₂H₃Cl₇