Order Entry
Norway
ContactUsLinkComponent
2,3,3',4,4',5,6-Heptachlorobiphenyl (PCB No. 190)
2,3,3',4,4',5,6-Heptachlorobiphenyl (PCB No. 190)
Catalog # PROOCIL-PCB-190-C
CAS Number:  
2,3,3',4,4',5,6-Heptachlorobiphenyl (PCB No. 190)
Catalog # PROOCIL-PCB-190-C
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :395.32 g/mol
CAS Number:41411-64-7
Formula:(C₆H₃Cl₂)(C₆Cl₅)