


α,α'-Dichloro-p-xylene 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:623-25-6
:
Formula:C₈H₈Cl₂
MDL Number:MFCD00000920
:175.06 g/mol
Boiling Pt:254 °C (760 mmHg)
Melting Pt:98…102 °C
Density:1,432 g/cm³ (20 °C)
EINECS:210-782-7
UN:2928
ADR:6.1,II
:GHS06
:GHS05
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to light yellow Crystalline solid
- Infrared spectrum:Conforms
- Melting point:98 to 102 °C
- GC:≥97.5 %