Phenolphthalein
Glass bottle for solids, 1 kg
Catalog # MOLE13440009-1KG
Marketsource
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one
- CAS Number:77-09-8
- Formula:C₂₀H₁₄O₄
- MDL Number:MFCD00005913
- Molecular Weight:318.33 g/mol
- Merck Index:14,07243
- Melting Pt:258…262 °C
- Density:1.3 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:201-004-7
- REACH:01-2119498295-24
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Cat. No.:MOLE13440009-1KG
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



