Order Entry
Norway
ContactUsLinkComponent
 
Irritant
Benzil
Catalog # EHERC10536300
Supplier:  EHRENSTORFER
CAS Number:  
Irritant
Benzil
Catalog # EHERC10536300
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :210.23 g/mol
Merck Index:13,01078
Boiling Pt:346 °C (1013 hPa)
Melting Pt:95 °C
Density:1.23 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:205-157-0
 :GHS07
CAS Number:134-81-6
Formula:(C₆H₅)C(O)C(O)(C₆H₅)
MDL Number:MFCD00003080