Bromocresol green ACS, Reag. Ph. Eur.
Supplier: Merck
Certificates
About this item
CAS Number:76-60-8
:
Formula:C₂₁H₁₄Br₄O₅S
MDL Number:MFCD00005874
:698.02 g/mol
Merck Index:13,01370
Melting Pt:217…218 °C
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:200-972-8
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:217 - 218 °C
- Molar Mass:698.02 g/mol
- Bulk Density:350 kg/m3
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC1.08121.0025_TDS_EN ProductPage_108121
Product specification
Merck Brighter titration Titripur®
Product Literature
msds_108121