kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
Bromocresol green
Bromocresol green
Bromocresol green
Bromocresol green
Supplier:
CHROMA
Certificates
Utvid bildet
Bromocresol green
Supplier:
CHROMA
About this item
CAS Number:
76-60-8
:
Formula:
C₂₁H₁₄Br₄O₅S
MDL Number:
MFCD00005874
:
698.02 g/mol
Merck Index:
13,01370
Melting Pt:
217…218 °C
Storage Temperature:
Ambient
EINECS:
200-972-8