Order Entry
Norway
ContactUsLinkComponent
Bromocresol green, pure indicator grade
Bromocresol green, pure indicator grade
Certificates
undefined
Bromocresol green, pure indicator grade

Specification Test Results

  • Appearance:
    Yellow to beige to light brown Powder
  • Infrared spectrum:
    Conforms
  • Solubility:
    (0.1 % in ethanol) Clear
  • UV Lambda max:
    613 to 621 nm (in buffer)
  • Specific extinction E (1%/1cm):
    ≥550 (at lambda max)
  • Visual transition interval:
    from pH 3.8 (yellow) to pH 5.4 (blue)
  • Heavy metals (as Pb):
    ≤10 ppm

Specification Test Results

Related Information