Order Entry
Norway
ContactUsLinkComponent
Benzyl thiocyanate
Benzyl thiocyanate
Catalog # MAYBJFD03762.1
Supplier:  Thermo Scientific
CAS Number:  
undefined
Benzyl thiocyanate
Catalog # MAYBJFD03762.1
Supplier:  Thermo Scientific
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :149.22 g/mol
Boiling Pt:230 °C (1013 hPa)
Melting Pt:38…41 °C
Density:1.32 g/cm³ (20 °C)
Flash Pt:>110 °C
Storage Temperature:Refrigerator
EINECS:221-144-2
UN:2811
ADR:6.1,III
CAS Number:3012-37-1
Formula:C₈H₇NS
MDL Number:MFCD00001832