Order Entry
Norway
ContactUsLinkComponent
4-Nitro-o-toluidine 99%
Toxic
Health
Environment
4-Nitro-o-toluidine 99%
Catalog # ACRO163950050
CAS Number:  
undefined
Toxic
Health
Environment
4-Nitro-o-toluidine 99%
Catalog # ACRO163950050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Yellow to brown to khaki Crystalline powder or needles
  • Infrared spectrum:
    Conforms
  • Melting point:
    129 to 133 °C
  • GC:
    ≥98.5 %

Specification Test Results

About this item

 :
 :152.15 g/mol
Melting Pt:129…133 °C
Density:1,366 g/cm³ (15 °C)
Storage Temperature:Ambient
EINECS:202-762-1
UN:2660
ADR:6.1,III
 :GHS06
 :GHS08
 :GHS09
CAS Number:99-52-5
Formula:C₇H₈N₂O₂
MDL Number:MFCD00007734