Order Entry
Norway
ContactUsLinkComponent
Phenethylamine for synthesis, Sigma-Aldrich®
Toxic
Corrosive
Phenethylamine for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Toxic
Corrosive
Phenethylamine for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    4.3 g/l (20 °C)
  • Molar Mass:
    121.18 g/mol
  • Boiling Point:
    200 - 202 °C (1013 hPa)
  • Vapor Pressure:
    0.4 hPa (20 °C)
  • Flash Point:
    81 °C
  • Refractive Index:
    1.5332 (20 °C, 589 nm)
  • Explosion Limit:
    1.0 - 5.5 %(V)
  • Density:
    0.96 g/cm3 (20 °C)
  • pH:
    11.5 (4.3 g/l, H2O, 20 °C)
  • Ignition Point:
    425 °C
  • Viscosity kinematic:
    2.40 mm2/s (23 °C)

Specification Test Results

Related Information