kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
4-Chloro-m-toluidine
4-Chloro-m-toluidine 98+%
4-Chloro-m-toluidine 98+%
:
Thermo Fisher Scientific
4-Chloro-m-toluidine 98+%
:
Thermo Fisher Scientific
:
7149-75-9
:
:
C₇H₈ClN
:
MFCD00066332
:
141.6 g/mol
:
241 °C (1013 hPa)
:
83…84 °C
:
2239
:
6.1,III
:
GHS07
Appearance:
Yellow Powder
Infrared spectrum:
Conforms
Melting point:
83 to 84 °C
GC:
≥98.0 %
ACRO38368