
2-Chlorobenzylamine 97%
:89-97-4
:
:C₇H₈ClN
:MFCD00008108
:141.6 g/mol
:103…104 °C (11 mmHg)
:1.17 g/cm³
:88 °C
:Ambient
:201-955-8
:2735
:8,II
:GHS05
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5610 to 1.5640 (20°C, 589 nm)
- GC:≥96.0 %