
4-Chloro-3-(trifluoromethyl)aniline 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:320-51-4
:
Formula:C₇H₅ClF₃N
MDL Number:MFCD00007846
:195.57 g/mol
Boiling Pt:85 °C (3 mmHg)
Melting Pt:34…38 °C
Flash Pt:134 °C
Storage Temperature:Ambient
EINECS:206-277-6
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:White to pink to light orange Crystalline powder or low melting solid
- Infrared spectrum:Conforms
- Melting point:34 to 38 °C
- GC:≥98.5 %