
2,5-Dichloro-α,α,α-trifluorotoluene ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:320-50-3
:
Formula:C₇H₃Cl₂F₃
MDL Number:MFCD00000608
:215 g/mol
Boiling Pt:181 °C (1013 hPa)
Melting Pt:11 °C
Density:1.483
Flash Pt:76 °C (168 °F)
Storage Temperature:Ambient
EINECS:206-276-0
UN:1760
ADR:8,III
:GHS05