Order Entry
Norway
Orders LinkContactUsLinkComponent
4'-Bromoacetophenone

4'-Bromoacetophenone

Glass bottle for solids, 100 g
Catalog # MOLE27065743-100G
Irritant
Marketsource
Supplier:   Molekula
4'-Bromoacetophenone
Catalog # MOLE27065743-100G
Supplier:  Molekula
Marketsource
CAS Number:  99-90-1
4'-Bromoacetophenone

Glass bottle for solids, 100 g

CAS Number:  99-90-1
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  p-Bromoacetophenone, 1-(4-Bromophenyl)ethanone
  • CAS Number:99-90-1
  • Formula:C₈H₇BrO
  • MDL Number:MFCD00000105
  • Molecular Weight:199.05 g/mol
  • Merck Index:13,01387
  • Boiling Pt:255 °C (1013 hPa)
  • Melting Pt:49…52 °C
  • Density:1.736 g/cm³ (20 °C)
  • Flash Pt:>100 °C
  • Storage Temperature:Ambient
  • EINECS:202-799-3
  • UN:2811
  • ADR:6.1,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.