4'-Bromoacetophenone ≥98%
Purity: 98% CAS No.: 99-90-1 Molecular Formula: C8H7BrO Formula Weight: 199.05 Flash Point: 110°(230°F) Melting Point: 49-53° Boiling Point: 255-256° Beilstein Reference: 386015 Merck Reference: 14,1403 MDL No.: MFCD00000105
: p-Bromoacetophenone, 1-(4-Bromophenyl)ethanone
- :99-90-1
- :C₈H₇BrO
- :MFCD00000105
- :199.05 g/mol
- :14,1403
- :255…256 °C
- :49…53 °C
- :1.736 g/cm³ (20 °C)
- :>110 °C (230 °F)
- :Ambient
- :202-799-3
- :2811
- :6.1,II
Purity: 98% CAS No.: 99-90-1 Molecular Formula: C8H7BrO Formula Weight: 199.05 Flash Point: 110°(230°F) Melting Point: 49-53° Boiling Point: 255-256° Beilstein Reference: 386015 Merck Reference: 14,1403 MDL No.: MFCD00000105
: p-Bromoacetophenone, 1-(4-Bromophenyl)ethanone
- :99-90-1
- :C₈H₇BrO
- :MFCD00000105
- :199.05 g/mol
- :14,1403
- :255…256 °C
- :49…53 °C
- :1.736 g/cm³ (20 °C)
- :>110 °C (230 °F)
- :Ambient
- :202-799-3
- :2811
- :6.1,II
Product Family Options
- Pack typePk