Order Entry
Norway
Orders LinkContactUsLinkComponent
 

4'-Bromoacetophenone 98%

Irritant
4'-Bromoacetophenone 98%
Certificates
4'-Bromoacetophenone 98%

3 Options Available Below

Product Details & Documents
Synonyms:  p-Bromoacetophenone, 1-(4-Bromophenyl)ethanone
  • CAS Number:99-90-1
  • Formula:C₈H₇BrO
  • MDL Number:MFCD00000105
  • Molecular Weight:199.05 g/mol
  • Merck Index:13,01387
  • Boiling Pt:255 °C (760 mmHg)
  • Melting Pt:48…52 °C
  • Density:1,736 g/cm³ (20 °C)
  • Flash Pt:>100 °C
  • Storage Temperature:Ambient
  • EINECS:202-799-3
  • ADR:6.1,III

 

Specifications for Product Family

Specification Test Results

  • Appearance:
    White to light yellow Crystals or flakes
  • Infrared spectrum:
    Conforms
  • Melting point:
    48 to 52 °C
  • GC:
    ≥97.5 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - kr 385,00
    kr 385,00
    1 * 25 g
       kr 385,00

  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - kr 935,00
    kr 935,00
    1 * 100 g
       kr 935,00

  • Glass bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - kr 3 275,00
    kr 3 275,00
    1 * 500 g
       kr 3 275,00