Order Entry
Norway
ContactUsLinkComponent
 
Anisole 99%, Reagent Grade
    
 
undefined
Anisole 99%, Reagent Grade
    

 

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion

 

 

  • Vapor Density:
    3.7 (vs air)
  • Assay (GC):
    Min. 98.5 %
  • Identity (IR):
    complying
  • Vapor Pressure:
    10 mmHg ( 42.2 °C)
  • Autoignition Temperature:
    887 °F