
4-Nitrobenzenesulphonyl chloride 95%
:98-74-8
:
:C₆H₄ClNO₄S
:MFCD00007445
:221.62 g/mol
:143…144 °C (2hPa)
:75…80 °C
:1,69 g/cm³ (20 °C)
:Ambient
:202-697-9
:3261
:8,II
:GHS05
- Appearance:Yellow to brown Powder
- Infrared spectrum:Conforms
- Melting point:75 to 80 °C
- GC:≥94 %