

3-Aminophenol
Catalog # MOLE12674641-500G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
Merck Index:13,00459
Boiling Pt:299 °C (1013 hPa)
Melting Pt:122 °C
Density:1.275 g/cm³ (20 °C)
Flash Pt:92 °C
EINECS:209-711-2
UN:2512
ADR:6.1,III
:109.13 g/mol
:GHS07
:GHS09
CAS Number:591-27-5
Formula:C₆H₇NO
MDL Number:MFCD00007786