Order Entry
Norway
ContactUsLinkComponent
 
 
 
3-Aminophenol for synthesis, Sigma-Aldrich®
 :  Merck
 
 
 
3-Aminophenol for synthesis, Sigma-Aldrich®
 :  Merck

 

  • Solubility:
    26 g/l (20 °C)
  • Melting Point:
    122 °C
  • Molar Mass:
    109.13 g/mol
  • Bulk Density:
    700 kg/m3
  • Boiling Point:
    164 °C (15 hPa)
  • Vapor Pressure:
     - 0.1 hPa (20 °C)
  • Flash Point:
    178 °C
  • Density:
    0.99 g/cm3 (25 °C)
  • pH:
    6.8 (10 g/l, H2O, 20 °C)
  • Ignition Point:
    600 °C
  • Synonyms:
    3-Hydroxyaniline
  • Hill Formula:
    C6H7NO
  • Chemical Formula:
    3-(NH2)C6H4OH