Order Entry
Norway
ContactUsLinkComponent
2-Aminophenol 99%
Irritant
Health
2-Aminophenol 99%
Certificates
undefined
Irritant
Health
2-Aminophenol 99%

Specification Test Results

  • Appearance (Color):
    White to brown
  • Appearance (Form):
    Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    172 to 177 °C
  • GC:
    ≥98.5 %

Specification Test Results

Related Information