Order Entry
Norway
ContactUsLinkComponent
3,5-Difluorobenzenesulphonyl chloride ≥97%
Corrosive
3,5-Difluorobenzenesulphonyl chloride ≥97%
Catalog # L19507.06
CAS Number:  
Corrosive
3,5-Difluorobenzenesulphonyl chloride ≥97%
Catalog # L19507.06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

About this item

CAS Number:210532-25-5
 :
Formula:C₆H₃ClF₂O₂S
MDL Number:MFCD02091378
 :212.6 g/mol
Melting Pt:57…59 °C
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
UN:3261
ADR:8,II
 :GHS05