Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Chlor-4-fluorothiophenol
2-Chlor-4-fluorothiophenol ≥97%
2-Chlor-4-fluorothiophenol ≥97%
:
Thermo Fisher Scientific
2-Chlor-4-fluorothiophenol ≥97%
:
Thermo Fisher Scientific
:
175277-99-3
:
:
C₆H₄ClFS
:
MFCD00052936
:
162.61 g/mol
:
86…88 °C (15 mmHg)
:
GHS06
:
GHS07
:
Ambient
msds_ALFAA13498
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years