Order Entry
Norway
ContactUsLinkComponent
4-(Trifluoromethoxy)benzenesulphonyl chloride
4-(Trifluoromethoxy)benzenesulphonyl chloride
Catalog # MAYBTL00343.1
Supplier:  Thermo Scientific
CAS Number:  
4-(Trifluoromethoxy)benzenesulphonyl chloride
Catalog # MAYBTL00343.1
Supplier:  Thermo Scientific
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :260.62 g/mol
Boiling Pt:120…122 °C (15 mmHg)
Density:1.536 g/cm³ (20 °C)
EINECS:302-397-9
UN:3265
ADR:8,II
CAS Number:94108-56-2
Formula:C₇H₄ClF₃O₃S
MDL Number:MFCD00042408