Order Entry
Norway
ContactUsLinkComponent
 
 
3,4,5-Trifluorobenzoic acid ≥98.0% (by titrimetric analysis)
 :  TCI
 
undefined
 
3,4,5-Trifluorobenzoic acid ≥98.0% (by titrimetric analysis)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    97.0 to 99.0 deg-C
  • Solubility in Methanol:
    very faint turbidity