Order Entry
Norway
ContactUsLinkComponent
2-Fluorobenzhydrazide
2-Fluorobenzhydrazide
Catalog # MAYBSPB03430.1
Supplier:  Thermo Scientific
CAS Number:  
2-Fluorobenzhydrazide
Catalog # MAYBSPB03430.1
Supplier:  Thermo Scientific
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :154.14 g/mol
CAS Number:446-24-2
Formula:C₇H₇FN₂O
MDL Number:MFCD00025112