4-(Trifluoromethoxy)benzonitrile
Supplier: Thermo Scientific
Certificates
About this item
CAS Number:332-25-2
:
Formula:C₈H₄F₃NO
MDL Number:MFCD00039474
Boiling Pt:192 °C (1013 hPa)
Density:1.284 g/cm³ (20 °C)
EINECS:206-363-3
UN:3276
ADR:6.1,III
:187.12 g/mol