Cobalt(II) benzoate
Supplier: Thermo Fisher Scientific
Jump to:
About this item
CAS No.: 932-69-4 Molecular Formula: C14H10CoO4 Formula Weight: 301.16 Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: Grey Odour: Odourless Melting Point: 114-116° MDL No.: MFCD00050769
Synonyms: Benzoic acid cobalt (II) salt, Cobalt dibenzoate, Cobaltous benzoate
- CAS Number:932-69-4
- Formula:C₁₄H₁₀CoO₄
- MDL Number:MFCD00050769
- Molecular Weight:301.16 g/mol
- Melting Pt:114…116 °C
- Storage Temperature:Ambient
- EINECS:213-257-0
2 Options Available Below
Product Details & Documents
CAS No.: 932-69-4 Molecular Formula: C14H10CoO4 Formula Weight: 301.16 Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: Grey Odour: Odourless Melting Point: 114-116° MDL No.: MFCD00050769
Synonyms: Benzoic acid cobalt (II) salt, Cobalt dibenzoate, Cobaltous benzoate
- CAS Number:932-69-4
- Formula:C₁₄H₁₀CoO₄
- MDL Number:MFCD00050769
- Molecular Weight:301.16 g/mol
- Melting Pt:114…116 °C
- Storage Temperature:Ambient
- EINECS:213-257-0
Documents
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



