Benzocaine (ethyl-4-aminobenzoate)
Catalog # EHERC10171450
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:165.19 g/mol
Merck Index:13,01087
Boiling Pt:311 °C (1013 hPa)
Melting Pt:89…90 °C
Flash Pt:112 °C
Storage Temperature:Ambient
EINECS:202-303-5
:GHS07
CAS Number:94-09-7
Formula:C₉H₁₁NO₂
MDL Number:MFCD00007892