

Tecnazene, Supelco®
Catalog # 45672-250MG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
Boiling Pt:304 °C (1013 hPa)
Melting Pt:99 °C
Density:1.744 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:3077
ADR:9,III
:M-Clarity™ quality level = MQ100
:260.89 g/mol
EINECS:204-178-2
:GHS07
:GHS09
CAS Number:117-18-0
Formula:C₆HCl₄NO₂
MDL Number:MFCD00007066