3-Chloro-2-fluorobenzamide
Catalog # MAYBCD11825.1
Supplier: Thermo Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:173.57 g/mol
CAS Number:104326-94-5
Formula:C₇H₅ClFNO
MDL Number:MFCD00042205