Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3-Dichlorobenzamide
2,3-Dichlorobenzamide
2,3-Dichlorobenzamide
MAYBTL00592.50
:
Thermo Scientific
:
2,3-Dichlorobenzamide
MAYBTL00592.50
:
Thermo Scientific
:
Pack type:
Glass bottle for solids
Pk:
50 g
Seq:
3368
:
:
190.03 g/mol
:
5980-24-5
:
C₇H₅Cl₂NO
:
MFCD00173938