
2,4,6-Trichlorobenzoyl chloride 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4136-95-2
:
Formula:C₇H₂Cl₄O
MDL Number:MFCD00075323
:243.9 g/mol
Boiling Pt:107…108 °C (6 mmHg)
Density:1.56 g/cm³
Flash Pt:>110 °C
Storage Temperature:Ambient
UN:3265
ADR:8,II
:GHS05
Specification Test Results
- Appearance:Clear yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5750 to 1.5770 (20°C, 589 nm)
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together