Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3-Dichlorophenylboronic acid
2,3-Dichlorophenylboronic acid ≥98%
2,3-Dichlorophenylboronic acid ≥98%
Irritant
2,3-Dichlorophenylboronic acid ≥98%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
2,3-Dichlorophenylboronic acid ≥98%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
151169-74-3
:
Formula:
C₆H₅BCl₂O₂
MDL Number:
MFCD01075703
:
190.82 g/mol
Melting Pt:
233…242 °C
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAB22781
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation