Anti- Horse Polyclonal Antibody (Unconjugated)
Catalog # IMMRIR2398
Supplier: ImmunoReagents
Specifications
- Antibody type:Control
- Antigen symbol:Ig
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Horse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:10 mg
- Pk:10 mg
Specifications
About this item
Type:
Antigen:
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Horse
Isotype:
Reactivity: