Anti- Goat Polyclonal Antibody (Unconjugated)
Catalog # IMMRIR2396
Leverandør: ImmunoReagents
Spesifikasjoner
- Antibody type:Control
- Antigen symbol:Ig
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:10 mg
- Pk:10 mg
Spesifikasjoner
Om dette produktet
Type:
Antigen:
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: