Anti-IgG Rat Polyclonal Antibody (Unconjugated)
Catalog # IMMRIR2375
Supplier: ImmunoReagents
Specifications
- Antibody type:Control
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rat
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Western blot:Yes
- Form:Clear, colorless liquid, 0.2 µm filtered
- Storage temperature:Cool
- Concentration:> 4.5 mg/ml (E 1% at 280 nm = 13.0)
- Shipping temperature:2-8 °C
- Purification:Protein G
- Size:5 mg
- Pk:5 mg
Specifications
About this item
Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: