Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Isopropyl-3,8-dimethylazulene-1-carbaldehyde
5-Isopropyl-3,8-dimethylazulene-1-carbaldehyde
5-Isopropyl-3,8-dimethylazulene-1-carbaldehyde
5-Isopropyl-3,8-dimethylazulene-1-carbaldehyde
Supplier:
Thermo Scientific
Certificates
Expand image
5-Isopropyl-3,8-dimethylazulene-1-carbaldehyde
Supplier:
Thermo Scientific
About this item
CAS Number:
3331-47-3
:
Formula:
C₁₆H₁₈O
MDL Number:
MFCD00275723
:
226.32 g/mol