Order Entry
Norway
Orders LinkContactUsLinkComponent
Succinimide

Succinimide

Glass bottle for solids, 1 kg
Catalog # MOLE48731952-1KG
Marketsource
Supplier:   Molekula
Succinimide
Catalog # MOLE48731952-1KG
Supplier:  Molekula
Marketsource
CAS Number:  123-56-8
Succinimide

Glass bottle for solids, 1 kg

CAS Number:  123-56-8
Price
kr 2 650,00
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Pyrrolidine-2,5-dione
  • CAS Number:123-56-8
  • Formula:C₄H₅NO₂
  • MDL Number:MFCD00005495
  • Molecular Weight:99.09 g/mol
  • Merck Index:12,09040
  • Boiling Pt:285 °C (1013 hPa)
  • Melting Pt:121…124 °C
  • Density:1.418 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:204-635-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.